CAS RN 465-42-9
IUPAC Namn:
(2E,4E,6E,8E,10E,12E,14E,16E,18E)-1-[(1R,4S)-4-hydroxy-1,2,2-trimethylcyclopentyl]-19-[(4R)-4-hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl]-4,8,13,17-tetramethylnonadeca-2,4,6,8,10,12,14,16,18-nonaen-1-one
Synonymer:
paprika extract capsanthin/capsorubin capsanthin (substance) capsanthin
Molekylvikt (g/mol):
584.89
Molekylär sammansättning:
C40H56O3
InChi Key:
VYIRVAXUEZSDNC-RDJLEWNRSA-N
SMILES:
C\C(\C=C\C=C(/C)\C=C\C(=O)[C@]1(C)C[C@@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C
1
–
2
av
2
Resultat
1
Capsanthin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Capsanthin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.