CAS RN 5063-03-6
IUPAC Namn:
1-{bicyclo[2.2.1]hept-5-en-2-yl}ethan-1-one
Synonymer:
1-{bicyclo[2.2.1]hept-5-en-2-yl}ethanone
Molekylvikt (g/mol):
136.19
Molekylär sammansättning:
C9H12O
InChi Key:
NIMLCWCLVJRPFY-UHFFFAOYNA-N
SMILES:
CC(=O)C1CC2CC1C=C2
1
–
3
av
3
Resultat
1
5-Acetyl-2-norbornene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Acetyl-2-norbornene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Acetyl-2-norbornene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.