CAS RN 54060-41-2
IUPAC Namn:
{3-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]-2,2,5,5-tetramethylpyrrolidin-1-yl}oxidanyl
Synonymer:
3-[(2,5-dioxopyrrol-1-yl)methyl]-2,2,5,5-tetramethylpyrrolidin-1-yloxidanyl
Molekylvikt (g/mol):
251.31
Molekylär sammansättning:
C13H19N2O3
InChi Key:
FHTCUAVCPMMPNP-UHFFFAOYNA-N
SMILES:
CC1(C)CC(CN2C(=O)C=CC2=O)C(C)(C)N1[O]
1
–
3
av
3
Resultat
1
1-Oxyl-3-(maleimidomethyl)-2,2,5,5-tetramethyl-1-pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Oxyl-3-(maleimidomethyl)-2,2,5,5-tetramethyl-1-pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Oxyl-3-(maleimidomethyl)-2,2,5,5-tetramethyl-1-pyrrolidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.