CAS RN 564-20-5
IUPAC Namn:
3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan-2-one
Synonymer:
sclareolide
Molekylvikt (g/mol):
250.38
Molekylär sammansättning:
C16H26O2
InChi Key:
IMKJGXCIJJXALX-UHFFFAOYNA-N
SMILES:
CC12CCC3C(C)(C)CCCC3(C)C1CC(=O)O2
1
–
3
av
3
Resultat
1
Sclareolide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sclareolide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sclareolide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.