CAS RN 57075-34-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
5,6-Diamino-2-(2-propoxyphenyl)pyrimidin-4(3H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5,6-Diamino-2-(2-propoxyphenyl)pyrimidin-4(3H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5,6-Diamino-2-(2-propoxyphenyl)pyrimidin-4(3H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.