CAS RN 61884-74-0
IUPAC Namn:
hydrogen (3S)-3-aminohexanedioic acid chloride
Synonymer:
hydrogen (3S)-3-aminohexanedioic acid chloride
Molekylvikt (g/mol):
197.62
Molekylär sammansättning:
C6H12ClNO4
InChi Key:
XMUKSNPNAOIQKU-WCCKRBBISA-N
SMILES:
[H+].[Cl-].N[C@@H](CCC(O)=O)CC(O)=O
1
–
1
av
1
Resultat
1
L-beta-Homoglutamic Acid Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.