CAS RN 6222-35-1
IUPAC Namn:
cyclohexyl propanoate
Synonymer:
propionic acid cyclohexyl ester
Molekylvikt (g/mol):
156.23
Molekylär sammansättning:
C9H16O2
InChi Key:
MAMMVUWCKMOLSG-UHFFFAOYSA-N
SMILES:
CCC(=O)OC1CCCCC1
1
–
3
av
3
Resultat
1
Cyclohexyl Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cyclohexyl Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cyclohexyl Propionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.