CAS RN 6362-80-7
IUPAC Namn:
(4-methyl-4-phenylpent-1-en-2-yl)benzene
Synonymer:
(4-methyl-4-phenylpent-1-en-2-yl)benzene
Molekylvikt (g/mol):
236.36
Molekylär sammansättning:
C18H20
InChi Key:
ZOKCNEIWFQCSCM-UHFFFAOYSA-N
SMILES:
CC(C)(CC(=C)C1=CC=CC=C1)C1=CC=CC=C1
1
–
3
av
3
Resultat
1
Nofmer MSD, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Nofmer MSD, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Nofmer MSD, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.