CAS RN 64063-37-2
IUPAC Namn:
2,6-dichloro-3-methylaniline
Synonymer:
2,6-dichloro-3-methylaniline
Molekylvikt (g/mol):
176.04
Molekylär sammansättning:
C7H7Cl2N
InChi Key:
AYVZXDFCFQSWJD-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Cl)C(N)=C1Cl
1
–
2
av
2
Resultat
1
2,6-Dichloro-3-methylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Dichloro-3-methylaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.