CAS RN 68744-64-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
5-Trifluoromethyl-4H-1,2,4-triazole-3(2H)thione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Trifluoromethyl-4H-1,2,4-triazole-3(2H)thione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Trifluoromethyl-4H-1,2,4-triazole-3(2H)thione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.