CAS RN 6979-94-8
IUPAC Namn:
[3,4-bis(acetyloxy)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl acetate
Synonymer:
[3,4-bis(acetyloxy)-5-(2-amino-6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl acetate
Molekylvikt (g/mol):
409.36
Molekylär sammansättning:
C16H19N5O8
InChi Key:
ULXDFYDZZFYGIY-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C1NC(N)=NC2=O
1
–
1
av
1
Resultat
1
2’,3’,5’-Tri-O-acetyl Guanosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.