CAS RN 71772-25-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
[3aR-(3aAlpha,4Alpha,6aAlpha)]-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[3aR-(3aAlpha,4Alpha,6aAlpha)]-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
[3aR-(3aa,4a,6aa)]-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.