CAS RN 720666-74-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(BetaR,DeltaR,1S,2S)-1,2-Dihydro-Beta,Delta,6-trihydroxy-2-methyl-1-naphthaleneheptanoic Acid Sodium Salt (Pravastatin Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(BetaR,DeltaR,1S,2S)-1,2-Dihydro-Beta,Delta,6-trihydroxy-2-methyl-1-naphthaleneheptanoic Acid Sodium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.