CAS RN 7677-21-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
N,N,N’,N’-Tetramethylethane-1,2-diamine Dihydrochloride (1,2-Bis(dimethylamino)ethane Dihydrochloride, N,N,N’,N’-Tetramethylethylenediamine Dihydrochloride), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.