CAS RN 78850-37-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(3aR,4R,7aR)-3a,7a-Dihydro-2-methyl-4-[(1S,2R)-1,2,3-tris(acetyloxy)propyl]-4H-pyrano[3,4-d]oxazole-6-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3aR,4R,7aR)-3a,7a-Dihydro-2-methyl-4-[(1S,2R)-1,2,3-tris(acetyloxy)propyl]-4H-pyrano[3,4-d]oxazole-6-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3aR,4R,7aR)-3a,7a-Dihydro-2-methyl-4-[(1S,2R)-1,2,3-tris(acetyloxy)propyl]-4H-pyrano[3,4-d]oxazole-6-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.