CAS RN 80-44-4
IUPAC Namn:
butyl benzenesulfonate
Synonymer:
butyl benzenesulfonate
Molekylvikt (g/mol):
214.28
Molekylär sammansättning:
C10H14O3S
InChi Key:
NIKBCKTWWPVAIC-UHFFFAOYSA-N
SMILES:
CCCCOS(=O)(=O)C1=CC=CC=C1
1
–
2
av
2
Resultat
1
Butyl Benzenesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Butyl Benzenesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.