CAS RN 80639-95-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfinyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate (Fenticonazole Sulfoxide Nitrate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Fenticonazole Sulfoxide Nitric Acid Salt, TRC
CAS: 80639-95-8 Molekylvikt (g/mol): 534.41