CAS RN 938071-77-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.