CAS RN 93957-50-7
IUPAC Namn:
3-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]prop-2-enal
Synonymer:
3-[3-(4-fluorophenyl)-1-isopropylindol-2-yl]prop-2-enal
Molekylvikt (g/mol):
307.37
Molekylär sammansättning:
C20H18FNO
InChi Key:
DVWHSTKQJBIYCK-UHFFFAOYSA-N
SMILES:
CC(C)N1C(C=CC=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
1
–
4
av
4
Resultat
1
(E)-3-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)acrylaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(E)-3-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)acrylaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(E)-3-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)acrylaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(E)-3-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]prop-2-enal, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.