Learn More
1,1'-[[(1,1-Dimethyl-ethyl)imino]bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene)]]-bis(3,3-diethylurea), Mikromol™
Product Code. 17909184
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
17909184 25 mg 25mg
1 options
Product Code. 17909184 Supplier Mikromol Supplier No. MM0128.050025
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-[[(1,1-Dimethyl-ethyl)imino]bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene)]]-bis(3,3-diethylurea)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-[((1,1-Dimethyl-ethyl)imino]bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene))]-bis(3,3-diethylurea)
CAS 2731998-00-6
Analyte or Component Names 1,1'-[((1,1-Dimethyl-ethyl)imino]bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene))]-bis(3,3-diethylurea)
Shipping Condition Room Temperature
Molecular Formula C36 H55 N5 O8
Certifications/Compliance ISO 17025
Synonym Celiprolol Hydrochloride Imp. E (EP), Celiprolol Imp. E (EP), 1,1'-[[(1,1-Dimethyl-ethyl)imino]bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene)]]-bis(3,3-diethylurea), Celiprolol Hydrochloride Impurity E, Celiprolol Impurity E
InChI Formula InChI=1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11)
SMILES CCN(CC)C(=O)Nc1ccc(OCC(O)CN(CC(O)COc2ccc(NC(=O)N(CC)CC)cc2C(=O)C)C(C)(C)C)c(c1)C(=O)C
IUPAC Name 3-[3-acetyl-4-[3-[[3-[2-acetyl-4-(diethylcarbamoylamino)phenoxy]-2-hydroxypropyl]-tert-butylamino]-2-hydroxypropoxy]phenyl]-1,1-diethylurea
Molecular Weight (g/mol) 685.85
Quantity 25 mg
Formula Weight 685.4051 g/mol