Läs mer
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, TRC
Produktkod. 30008209
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30008209 1 g
1 options
Produktkod. 30008209 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2944001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
CAS 1515-14-6
Molekylformel C4 H4 F6 O
Kvantitet 1 g
Synonym 2-Propanol, 1,1,1,3,3,3-hexafluoro-2-methyl-, 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 1,1,1,3,3,3-Hexafluoro-2-hydroxy-2-methylpropane, 1,1,1-Trifluoro-2-trifluoromethyl-2-propanol, 1,1-Bis(trifluoromethyl)ethanol, 2-Methylhexafluoro-2-propanol, Hexafluoro-tert-butanol, Hexafluoro-tert-butyl alcohol, Perfluoro-(2-methyl)2-propanol
InChI formel InChI=1S/C4H4F6O/c1-2(11,3(5,6)7)4(8,9)10/h11H,1H3
LEDER CC(O)(C(F)(F)F)C(F)(F)F
IUPAC-namn 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
Molekylvikt (g/mol) 182.064
Formel vikt 182.0166