missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone trideuterate, 99.5% (Isotopic), Thermo Scientific™

Catalog Number 15466445
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.71 g/mL
Sensitivity Moisture
Quantity 10 g
CAS 109640-39-3
Melting Point 21°C
Molecular Formula C15H16F30O13
MDL Number MFCD09953659,MFCD09953659,MFCD00149064
UN Number UN2552
Synonym hexafluoroacetone deuterate 95.0atom%d
InChI Key MXXAOJTWEBEIPQ-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F
IUPAC Name pentakis(hexafluoropropan-2-one) octahydrate
Molecular Weight (g/mol) 974.23
PubChem CID 131844136
Formula Weight 226.10 (166.02 Anhydrous)
Percent Purity 99.5%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone trideuterate, Isotopic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'