Läs mer
1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol], Mikromol™
Produktkod. 30050926
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050926 25 mg
30051109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050926 Leverantör Mikromol Leverantörsnummer MM0082.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
CAS 130115-65-0
Analyt- eller komponentnamn 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol]
Frakt skick Room Temperature
Molekylformel C25 H31 N3 O4
Certifikat och efterlevnad ISO 9001
Synonym 1,1'-[(1-Methylethyl)imino]bis[3-(1H-indol-4-yloxy)propan-2-ol], Pindolol Imp. C (EP), Pindolol Impurity C
InChI formel InChI=1S/C25H31N3O4/c1-17(2)28(13-18(29)15-31-24-7-3-5-22-20(24)9-11-26-22)14-19(30)16-32-25-8-4-6-23-21(25)10-12-27-23/h3-12,17-19,26-27,29-30H,13-16H2,1-2H3
LEDER CC(C)N(CC(O)COc1cccc2[nH]ccc12)CC(O)COc3cccc4[nH]ccc34
IUPAC-namn 1-[[2-hydroxy-3-(1H-indol-4-yloxy)propyl]-propan-2-ylamino]-3-(1H-indol-4-yloxy)propan-2-ol
Molekylvikt (g/mol) 437.53
Kvantitet 25 mg
Formel vikt 437.2315 g/mol