Läs mer
1,1,1-Trichloro-2-methylpropan-2-ol, TRC
Produktkod. 30137938
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137938 10 g
30137151 5 g
17984475 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT73411010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57-15-8
Molekylformel C4 H7 Cl3 O
Molekylvikt (g/mol) 177.46
Synonym Chlorobutanol anhydrous, 1,1,1-Trichloro-2-methylpropan-2-ol, 1,1,1-Trichloro-tert-butyl alcohol, 2,2,2-Trichloro-1,1-dimethylethanol, 2-(Trichloromethyl)-2-propanol, Acetochlorone, Acetonchloroform, Acetone chloroform, Anhydrous chlorobutanol, Chlorbutanol, Chlorbutol, Chloreton, Chloretone, Chlorobutanol, Chlortran, Clortran, Coliquifilm, Dentalone, Khloreton, Methaform, NSC 44794, NSC 4596, NSC 5208, Sedaform, Trichloro-tert-butyl alcohol, Trichlorobutanol, β,β,β-Trichloro-tert-butyl alcohol
IUPAC-namn 1,1,1-trichloro-2-methylpropan-2-ol
LEDER CC(C)(O)C(Cl)(Cl)Cl
Specifikationer
Kemiskt namn eller material Chlorobutanol
Anteckningar om renhetsgrad GC
Kvantitet 10 g
InChI formel InChI=1S/C4H7Cl3O/c1-3(2,8)4(5,6)7/h8H,1-2H3
Formel vikt 175.9562
Procent renhet >95%