missing translation for 'onlineSavingsMsg'
Läs mer

1,1,1-Trifluoromethanesulfonic Acid (E)-4-[1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenyl Ester, TRC

Produktkod. 30450472
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450472 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30450472 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79055525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 1,1,1-Trifluoromethanesulfonic Acid (E)-4-[1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenyl Ester
CAS 1370699-80-1
Molekylformel C27H28F3NO4S
Kvantitet 25 mg
InChI formel InChI=1S/C27H28F3NO4S/c1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(15-11-21)34-19-18-31(2)3)22-12-16-24(17-13-22)35-36(32,33)27(28,29)30/h5-17H,4,18-19H2,1-3H3/b26-25+
LEDER CC\C(=C(\c1ccc(OCCN(C)C)cc1)/c2ccc(OS(=O)(=O)C(F)(F)F)cc2)\c3ccccc3
IUPAC-namn [4-[(E)-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenylbut-1-enyl]phenyl] trifluoromethanesulfonate
Molekylvikt (g/mol) 519.58
Formel vikt 519.1691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.