Läs mer
1,1,2-Trifluoro-2-(heptafluoropropoxy)ethene 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Produktkod. 30316066
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30316066 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 30316066 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA17846310AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 1 ml
Koncentration 100 μg/mL
Certifikat och efterlevnad ISO 17034
Lösningsmedel eller matris Acetonitrile