Läs mer
1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane, TRC
Produktkod. 30004783
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004783 500 mg
30008522 100 mg
30005809 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004783 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD535793500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 995-82-4
Molekylformel C12H38O5Si6
Molekylvikt (g/mol) 430.94
Synonym Hexasiloxane, 1,1,3,3,5,5,7,7,9,9,11,11-dodecamethyl- (6CI, 7CI, 8CI, 9CI, ACI), 1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane (ACI), 1,11-Dihydrododecamethylhexasiloxane, 1,11-Dihydrogendodecamethylhexasiloxane, Dodecamethyl-1,11-dihydrohexasiloxane
IUPAC-namn dimethylsilyloxy-[[[dimethylsilyloxy(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane
LEDER C[SiH](C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[SiH](C)C
Specifikationer
Kemiskt namn eller material 1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane
Kvantitet 500 mg
InChI formel InChI=1S/C12H38O5Si6/c1-18(2)13-20(5,6)15-22(9,10)17-23(11,12)16-21(7,8)14-19(3)4/h18-19H,1-12H3
Formel vikt 430.1335