Läs mer
1,1,3,3-Tetrachloroacetone (~85%), TRC
Produktkod. 30133620
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133620 2.5 g 2.5g
30008564 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2913402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 632-21-3
Molekylformel C3 H2 Cl4 O
Molekylvikt (g/mol) 195.86
Synonym 1,1,3,3-Tetrachloro-2-propanone (ACI), 1,1,3,3-Tetrachloroacetone, 1,1,3,3-Tetrachloropropanone;
IUPAC-namn 1,1,3,3-tetrachloropropan-2-one
LEDER ClC(Cl)C(=O)C(Cl)Cl
Specifikationer
Kemiskt namn eller material 1,1,3,3-Tetrachloroacetone
Kvantitet 2.5 g
InChI formel InChI=1S/C3H2Cl4O/c4-2(5)1(8)3(6)7/h2-3H
Formel vikt 193.886