missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%, Thermo Scientific™
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifikationer
Specifikationer
| Kokpunkt | 160°C to 161°C |
| Kemiskt namn eller material | 1,1,3,3-Tetramethyl-1,3-divinyldisilazane |
| Kvantitet | 100 g |
| Molekylformel | ClH |
| UN-nummer | UN2924 |
| Beilstein | 2411661 |
| Flampunkt | 42°C (107°F) |
| Synonym | deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide |
| Löslighetsinformation | Hydrolyzes in water. |
| InChI-nyckel | VEXZGXHMUGYJMC-FTGQXOHASA-N |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?