Läs mer
1-[1-(4-Chlorobenzoyl)-5-hydroxy-2-methyl-1H-indole-3-acetate]Beta-D-Glucopyranuronic Acid, TRC
Produktkod. 30363299
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30363299 5 mg
1 options
Produktkod. 30363299 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4291155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[1-(4-Chlorobenzoyl)-5-hydroxy-2-methyl-1H-indole-3-acetate]beta-D-Glucopyranuronic Acid
CAS 75511-22-7
Molekylformel C24 H22 Cl N O10
Kvantitet 5 mg
Synonym O-Desmethyl Indomethacin-acyl-β-D-glucuronide
InChI formel InChI=1S/C24H22ClNO10/c1-10-14(9-17(28)35-24-20(31)18(29)19(30)21(36-24)23(33)34)15-8-13(27)6-7-16(15)26(10)22(32)11-2-4-12(25)5-3-11/h2-8,18-21,24,27,29-31H,9H2,1H3,(H,33,34)/t18-,19-,20+,21-,24+/m0/s1
LEDER CC1=C(CC(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)=O)C3=CC(O)=CC=C3N1C(C4=CC=C(Cl)C=C4)=O
IUPAC-namn (2S,3S,4S,5R,6S)-6-[2-[1-(4-chlorobenzoyl)-5-hydroxy-2-methylindol-3-yl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 519.89