Läs mer
1,1-Bis-(4-Bromophenyl)-2,2-dichloroethylene, TRC
Produktkod. 30180000
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180000 50 mg
30179871 100 mg
30179864 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB55892050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21655-73-2
Molekylformel C14 H8 Br2 Cl2
Molekylvikt (g/mol) 406.93
Synonym 2,2-Bis(p-bromophenyl)-1,1-dichloroethene, p,p'-DDE dibromide, p,p'-DDBrE
IUPAC-namn 1-bromo-4-[1-(4-bromophenyl)-2,2-dichloroethenyl]benzene
LEDER ClC(=C(c1ccc(Br)cc1)c2ccc(Br)cc2)Cl
Specifications
Kemiskt namn eller material Dibromo-DDE
Kvantitet 50 mg
InChI formel InChI=1S/C14H8Br2Cl2/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8H
Formel vikt 403.837