Läs mer
1,1-Bis(4-chlorophenyl)ethane, Mikromol™
Produktkod. 30461717
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461717 4 x 25 mg
17991236 100 mg
17951896 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461717 Leverantör Mikromol Leverantörsnummer MM2513.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Bis(4-chlorophenyl)ethane, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1-Bis(4-chlorophenyl)ethane
CAS 4-4-3547
Analyt- eller komponentnamn 1,1-Bis(4-chlorophenyl)ethane
Frakt skick Room Temperature
Molekylformel C14 H12 Cl2
Certifikat och efterlevnad ISO 17025
Synonym 1,1-Bis(4-chlorophenyl)ethane
InChI formel InChI=1S/C14H12Cl2/c1-10(11-2-6-13(15)7-3-11)12-4-8-14(16)9-5-12/h2-10H,1H3
Lösningstyp Neat
LEDER CC(c1ccc(Cl)cc1)c2ccc(Cl)cc2
IUPAC-namn 1-chloro-4-[1-(4-chlorophenyl)ethyl]benzene
Molekylvikt (g/mol) 251.15
Kvantitet 4 x 25 mg
Formel vikt 250.0316 g/mol