Läs mer
1,1'-Carbonyldipiperidine, TRC
Produktkod. 30354218
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30354218 25 mg
1 options
Produktkod. 30354218 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17721025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1'-Carbonyldipiperidine
CAS 5395-04-0
Molekylformel C11H20N2O
Kvantitet 25 mg
Synonym Di-1-piperidinylmethanone, Bis(1-piperidyl) Ketone, Bis(pentamethylene)urea, Dipiperidin-1-ylmethanone, N,N,N',N'-Bis(cyclopentamethylene)urea, N,N,N',N'-Bispentamethyleneurea, N,N'-Carbonylbis(piperidine), NSC 3145
InChI formel InChI=1S/C11H20N2O/c14-11(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10H2
LEDER O=C(N1CCCCC1)N2CCCCC2
IUPAC-namn di(piperidin-1-yl)methanone
Molekylvikt (g/mol) 196.29
Formel vikt 196.1576