Läs mer
1,1-Dichloro-2-propanone, TRC
Produktkod. 30040379
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040379 10 mg
30005720 100 mg
30011419 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43547510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 513-88-2
Molekylformel C3 H4 Cl2 O
Molekylvikt (g/mol) 126.97
Synonym 1,1-Dichloro-2-propanone (ACI), 1,1-Dichloroacetone, 1,1-Dichloropropanone, Dichloromethyl methyl ketone, NSC 42725, α,α-Dichloroacetone
IUPAC-namn 1,1-dichloropropan-2-one
LEDER CC(=O)C(Cl)Cl
Specifikationer
Kemiskt namn eller material 1,1-Dichloropropanone-2
Kvantitet 10 mg
InChI formel InChI=1S/C3H4Cl2O/c1-2(6)3(4)5/h3H,1H3
Formel vikt 125.9639