Läs mer
1,1-Dichloroethane (Stabilized with Nitromethane, ~0.5%) (>90%), TRC
Produktkod. 30134815
Change view
Kvantitet:
1 g
2.5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134815 2.5 g
30133420 10 g
30010850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4342452.5G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75-34-3
Molekylformel C2 H4 Cl2
Molekylvikt (g/mol) 98.96
Synonym 1,1-Dichloroethane, 1,1-Ethylene dichloride, Dichloromethylmethane, Ethylidene chloride, Ethylidene dichloride, F 150a, HCC 150a
IUPAC-namn 1,1-dichloroethane
LEDER CC(Cl)Cl
Specifikationer
Kemiskt namn eller material 1,1-Dichloroethane
Kvantitet 2.5 g
InChI formel InChI=1S/C2H4Cl2/c1-2(3)4/h2H,1H3
Formel vikt 97.969