missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Diethyl-2,2'-cyanine iodide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1,1'-Diethyl-2,2'-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1′-Diethyl-2,2′-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).
Solubility
Soluble to 10 mM in dimethyl sulfoxide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
1,1′-Diethyl-2,2′-cyanine iodide is used to inhibit 1-methyl-4-phenylpyridinium (MPP+), toxic metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine that induces apoptosis in cerebellar granule neurons (CGNs).
Solubility
Soluble to 10 mM in dimethyl sulfoxide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications
Melting Point | 273°C (decomposition) |
Quantity | 1 g |
UN Number | UN2811 |
Sensitivity | Light sensitive |
Solubility Information | Soluble to 10 mM in dimethyl sulfoxide. |
Formula Weight | 454.35 |
Percent Purity | 97% |
Chemical Name or Material | 1,1'-Diethyl-2,2'-cyanine iodide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'