missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-4,4'-carbocyanine bromide, 96%, Thermo Scientific™

Catalog Number 15435207
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
19764-88-6
C25H25BrN2
433.393
MFCD06409816
QRRYUUHAKZXUIQ-UHFFFAOYSA-M
102601750
(4Z)-1-ethyl-4-[(E)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;bromide
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[Br-]
This item is not returnable. View return policy
19764-88-6
C25H25BrN2
433.393
MFCD06409816
QRRYUUHAKZXUIQ-UHFFFAOYSA-M
102601750
(4Z)-1-ethyl-4-[(E)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;bromide
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[Br-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 433.39
Percent Purity 96%
Chemical Name or Material 1,1'-Diethyl-4,4'-carbocyanine bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'