Läs mer
1,1-Dimethyl-3-cyanoguanidine, Mikromol™
Produktkod. 30052251
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052251 100 mg
30049273 25 mg
30458741 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30052251 Leverantör Mikromol Leverantörsnummer MM0020.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Dimethyl-3-cyanoguanidine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1-Dimethyl-3-cyanoguanidine
CAS 1609-06-9
Analyt- eller komponentnamn 1,1-Dimethyl-3-cyanoguanidine
Frakt skick Room Temperature
Molekylformel C4 H8 N4
Certifikat och efterlevnad ISO 17025
Synonym 1,1-Dimethyl-3-cyanoguanidine
InChI formel InChI=1S/C4H8N4/c1-8(2)4(6)7-3-5/h1-2H3,(H2,6,7)
LEDER CN(C)C(=N)NC#N
IUPAC-namn 3-cyano-1,1-dimethylguanidine
Molekylvikt (g/mol) 112.13
Kvantitet 100 mg
Formel vikt 112.0749 g/mol