missing translation for 'onlineSavingsMsg'
Läs mer

1,1-Dimethyl-d6-urea, CDN

Produktkod. 30326307
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30326307 0.25 g
30310670 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Reglering: Begränsningar kan gälla för köp av denna produkt och vi kan behöva begära ytterligare dokumentation från dig.
Produktkod. 30326307 Leverantör CDN Leverantörsnummer CDND62180.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N,N-Dimethylurea-D6
CAS 1219802-32-0
Molekylformel C3 D6 H2 N2 O
Kvantitet 0.25 g
Synonym N,N-Di(methyl-d3)urea, N,N-Dimethylurea-d6, N,N-Dimethylurea-D6, 1,1-Dimethyl-d6-urea, 1,1-Dimethylurea-d6, asym-Dimethylurea-d6, N,N-Di(methyl-d3)carbamide
InChI formel InChI=1 S/C3H8N2O/c1-5(2)3(4)6/h1-2H3,(H2,4,6)/i1D3,2D3
LEDER [2 H]C([2 H])([2 H])N(C(=O)N)C([2 H])([2 H])[2 H]
IUPAC-namn 1,1-bis(trideuteriomethyl)urea
Molekylvikt (g/mol) 94.1454
Formel vikt 94.1013 g/mol
Procent renhet 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.