missing translation for 'onlineSavingsMsg'
Läs mer

1-(1-Ethylpropyl)-5,6-dimethyl-7-nitro-1H-benzimidazole, TRC

Produktkod. 30136198
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136198 1 g
30135193 500 mg
30009117 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30136198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8329051G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 73215-11-9
Molekylformel C14 H19 N3 O2
Molekylvikt (g/mol) 261.3196
Synonym 1H-Benzimidazole, 1-(1-ethylpropyl)-5,6-dimethyl-7-nitro-, 1-(1-Ethylpropyl)-5,6-dimethyl-7-nitro-1H-benzimidazole
IUPAC-namn 5,6-dimethyl-7-nitro-1-pentan-3-ylbenzimidazole
LEDER CCC(CC)n1cnc2cc(C)c(C)c(c12)[N+](=O)[O-]

Specifikationer

Kemiskt namn eller material Pendimethalin metabolite 1 M455H029
Kvantitet 1 g
InChI formel InChI=1S/C14H19N3O2/c1-5-11(6-2)16-8-15-12-7-9(3)10(4)13(14(12)16)17(18)19/h7-8,11H,5-6H2,1-4H3
Formel vikt 261.1477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.