Läs mer
1,1'-Ferrocenedicarboxylic Acid, TRC
Produktkod. 30180328
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180328 250 mg
30180093 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180328 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF307620250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1293-87-4
Molekylformel C12H10FeO4
Molekylvikt (g/mol) 274.05
Synonym 1,1'-Dicarboxyferrocene, 1,1'-Bis(carboxy)ferrocene, 1,1'-Biscarboxylferrocene, Ferrocenedicarboxylic Acid;
IUPAC-namn 1,1′-Dicarboxyferrocene
LEDER O=C(O)[C-]12[CH]3=[CH]4[CH]5=[CH]1[Fe+2]45678932[CH]=%10[CH]9=[CH]8[C-]7(C(=O)O)[CH]%106
Specifikationer
Kemiskt namn eller material 1,1'-Ferrocenedicarboxylic Acid
Kvantitet 250 mg
InChI formel InChI=1S/2C6H5O2.Fe/c2*7-6(8)5-3-1-2-4-5;/h2*1-4H,(H,7,8);/q2*-1;+2
Formel vikt 273.99