Läs mer
1,1-Methanediyl Bismethanethiosulfonate, TRC
Produktkod. 30136943
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136943 5 mg
30135123 25 mg
30003635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2588005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22418-52-6
Molekylformel C3 H8 O4 S4
Molekylvikt (g/mol) 236.35
IUPAC-namn bis(methylsulfonylsulfanyl)methane
LEDER CS(=O)(=O)SCSS(=O)(=O)C
Specifikationer
Kemiskt namn eller material 1,1-Methanediyl Bismethanethiosulfonate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C3H8O4S4/c1-10(4,5)8-3-9-11(2,6)7/h3H2,1-2H3
Formel vikt 235.9305
Procent renhet >95%