Läs mer
1,1’-Methylene Bis[Theobromine], TRC
Produktkod. 30478681
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30478681 100 mg
1 options
Produktkod. 30478681 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303945100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1'-Methylene Bis[Theobromine]
CAS 77196-87-3
Molekylformel C15H16N8O4
Kvantitet 100 mg
Synonym 1,1'-Methylenebis[3,7-dihydro-3,7-dimethyl-1H-purine-2,6-dione, Pentoxifylline EP Impurity E
InChI formel InChI=1S/C15H16N8O4/c1-18-5-16-10-8(18)12(24)22(14(26)20(10)3)7-23-13(25)9-11(17-6-19(9)2)21(4)15(23)27/h5-6H,7H2,1-4H3
LEDER CN1C=NC2=C1C(=O)N(C(=O)N2C)CN3C(=O)C4=C(N=CN4C)N(C3=O)C
Molekylvikt (g/mol) 372.34