Läs mer
1-[(1-Oxopentyl)amino]-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-cyclopentanecarboxamide, TRC
Produktkod. 18010810
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18010810 5 mg
18093600 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18010810 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8599805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 748812-53-5
Molekylformel C25 H30 N6 O2
Molekylvikt (g/mol) 446.54
Synonym 1-(Pentanoylamino)-N-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]cyclopentanecarboxamide, Irbesartan Imp. A (EP), Irbesartan Related Compound A (USP), 1-[(1-Oxopentyl)amino]-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-cyclopentanecarboxamide
IUPAC-namn 1-(pentanoylamino)-N-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]cyclopentane-1-carboxamide
LEDER CCCCC(=O)NC1(CCCC1)C(=O)NCc2ccc(cc2)c3ccccc3c4nnn[nH]4
Specifikationer
Kemiskt namn eller material 1-(Pentanoylamino)-N-[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]cyclopentanecarboxamide
Kvantitet 5 mg
InChI formel InChI=1S/C25H30N6O2/c1-2-3-10-22(32)27-25(15-6-7-16-25)24(33)26-17-18-11-13-19(14-12-18)20-8-4-5-9-21(20)23-28-30-31-29-23/h4-5,8-9,11-14H,2-3,6-7,10,15-17H2,1H3,(H,26,33)(H,27,32)(H,28,29,30,31)
Formel vikt 446.243