Läs mer
1,10-Phenanthrolin-5-amine, TRC
Produktkod. 30420740
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420740 1 g
30409645 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420740 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2950301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,10-Phenanthrolin-5-amine
CAS 54258-41-2
Molekylformel C12 H9 N3
Kvantitet 1 g
Synonym 5-Amino-1,10-phenanthroline, 5-Amino-o-phenanthroline, 5-Aminophenanthroline
InChI formel InChI=1S/C12H9N3/c13-10-7-8-3-1-5-14-11(8)12-9(10)4-2-6-15-12/h1-7H,13H2
LEDER Nc1cc2cccnc2c3ncccc13
IUPAC-namn 1,10-phenanthrolin-5-amine
Molekylvikt (g/mol) 195.22
Formel vikt 195.0796