Läs mer
1,11-Diamino-3,6,9-trioxaundecane, TRC
Produktkod. 30350596
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350596 50 mg
30355011 100 mg
30356859 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30350596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41749350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 929-75-9
Molekylformel C8H20N2O3
Molekylvikt (g/mol) 192.26
IUPAC-namn 2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethanamine
LEDER NCCOCCOCCOCCN
Specifikationer
Kemiskt namn eller material 1,11-Diamino-3,6,9-trioxaundecane
Kvantitet 50 mg
InChI formel InChI=1S/C8H20N2O3/c9-1-3-11-5-7-13-8-6-12-4-2-10/h1-10H2
Formel vikt 192.1474