missing translation for 'onlineSavingsMsg'
Läs mer

1,12-Dodecanediol, TRC

Produktkod. 30375441
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375441 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375441 Supplier Toronto Research Chemicals Supplier No. TRCD494735100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1,12-Dodecanediol
CAS 5675-51-4
Molekylformel C12 H26 O2
Kvantitet 100 g
Synonym 1,12-Dodecanediol (6CI, 7CI, 8CI, 9CI, ACI), 1,12-Dihydroxydodecane, D 0969, Dodecamethylene glycol, n-Dodecane-1,12-diol, NSC 81250, α,ω-Dodecanediol
InChI formel InChI=1S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14H,1-12H2
LEDER OCCCCCCCCCCCCO
IUPAC-namn dodecane-1,12-diol
Molekylvikt (g/mol) 202.33
Formel vikt 202.1933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.