Learn More
Trajan Scientific and Medical™ 1/16 in. Graphite/Vespel™ Fittings Reducing Ferrule
Made from 15% Graphite and 85% Vespel
685.00 SEK - 900.00 SEK
Specifications
Material | 15% Graphite, 85% Vespel |
---|---|
Length (Metric) | 3.7 mm |
Width (Metric) | 3.2 mm |
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
12786348
|
Trajan Scientific and Medical™
0726632 |
0.2 mm | 3.7 mm | 3.2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15268883
|
Trajan Scientific and Medical™
072656 |
1.0 mm | 3.7 mm | 3.2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12766348
|
Trajan Scientific and Medical™
072658 |
0.9 mm | 3.7 mm | 3.2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms strong grip with capillary column
- Hole I.D. smaller than nominal size of fitting
Specifications
15% Graphite, 85% Vespel | |
3.7 mm | |
3.2 mm |