Learn More
Trajan Scientific and Medical™ 1/16 in. Male/Female Connectors and Ferrules
Forms an integral part of GC system.
580.00 SEK - 755.00 SEK
Specifications
Type | Connector |
---|---|
Diameter (English) Outer | 1/16 in. |
Material | Stainless Steel |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15249123
|
Trajan Scientific and Medical™
1034604 |
Male-Female Connector and Ferrules |
580.00 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15259123
|
Trajan Scientific and Medical™
1034605 |
Male-Female Tee Connector and Ferrules |
755.00 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
Connector | |
1/16 in. | |
Stainless Steel |