Läs mer
1-[(1R)-1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-1,2-1,2-bis(1,1-dimethylethyl) Ester Hydrazinedicarboxylic Acid, TRC
Produktkod. 30347517
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30347517 25 mg
1 options
Produktkod. 30347517 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB43186925MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-[(1R)-1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-1,2-1,2-bis(1,1-dimethylethyl) Ester Hydrazinedicarboxylic Acid
CAS 1281895-47-3
Molekylformel C27H42N2O9
Kvantitet 25 mg
Synonym 1-[(1R)-1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-1,2-1,2-bis(1,1-dimethylethyl) Ester Hydrazinedicarboxylic Acid, 1,2-Bis(1,1-dimethylethyl) 1-[(1R)-1-(3,4-dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-1,2-hydrazinedicarboxylate
InChI formel InChI=1S/C27H42N2O9/c1-12-18(13-2)36-22(31)27(9,21(30)17-14-15-19(34-10)20(16-17)35-11)29(24(33)38-26(6,7)8)28-23(32)37-25(3,4)5/h14-16,18H,12-13H2,1-11H3,(H,28,32)/t27-/m1/s1
LEDER CCC(CC)OC(=O)[C@](C)(N(NC(=O)OC(C)(C)C)C(=O)OC(C)(C)C)C(=O)c1ccc(OC)c(OC)c1
IUPAC-namn pentan-3-yl (2R)-3-(3,4-dimethoxyphenyl)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonyl-[(2-methylpropan-2-yl)oxycarbonylamino]amino]-3-oxopropanoate
Molekylvikt (g/mol) 538.64
Formel vikt 538.289